Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92848

Supelco

Trioctylphosphine oxide

for ion-selective electrodes, Selectophore

Synonym(s):

Thorium ionophore I, (Oct)3PO, TOPO®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]3PO
CAS Number:
Molecular Weight:
386.63
Beilstein/REAXYS Number:
1796648
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

Quality Level

product line

Selectophore

form

solid

bp

201-202 °C/2 mmHg (lit.)

mp

50-52 °C (lit.)

SMILES string

CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

InChI

1S/C24H51OP/c1-4-7-10-13-16-19-22-26(25,23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

ZMBHCYHQLYEYDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
TOPO is a registered trademark of Life Technologies

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

446.0 °F - closed cup

flash_point_c

230 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harikrishna Bavireddi et al.
The Analyst, 137(21), 5123-5127 (2012-09-25)
Highly fluorescent water soluble glyco-quantum dots were synthesized using a sonochemical procedure. The synthetic approach is based on specific host-guest interactions between β-cyclodextrin (β-CD) and trioctylphosphine oxide (TOPO) surfactant on quantum dots. The modified QDs were analyzed by a combination
Jungkon Kim et al.
Aquatic toxicology (Amsterdam, Netherlands), 97(2), 116-124 (2010-01-20)
The potential ecotoxicological consequences about semiconductor crystal nanoparticles (NPs) are a growing concern. However, our understanding of the mechanism of toxicity in NPs is very limited, especially under varying environmental conditions such as ultraviolet (UV) light. We performed an in
Yong Tao et al.
Journal of chromatography. A, 1216(35), 6259-6266 (2009-07-28)
By using ionic liquid as membrane liquid and tri-n-octylphosphine oxide (TOPO) as additive, hollow fiber supported liquid phase microextraction (HF-LPME) was developed for the determination of five sulfonamides in environmental water samples by high-performance liquid chromatography with ultraviolet detection The
Wesley E Smith et al.
ACS nano, 6(11), 9475-9484 (2012-10-09)
Semiconductor quantum dots (Qdots) are a promising new technology with benefits in the areas of medical diagnostics and therapeutics. Qdots generally consist of a semiconductor core, capping shell, and surface coating. The semiconductor core of Qdots is often composed of
Shuang-Yuan Zhang et al.
Advanced materials (Deerfield Beach, Fla.), 24(32), 4369-4375 (2012-07-19)
The preparation of vertically or horizontally aligned self-assemblies of CoP nanowires is demonstrated for the first time by aging them in the reaction solution for a sufficient time at 20 or 0 °C. This strategy opens up a way for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service