Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92804

Supelco

4-[2-(tert-Butylamino)-1-methoxyethyl]-2-(hydroxymethyl)phenol

pharmaceutical impurity standard

Synonym(s):

Salbutamol impurity A (PhEur), 5-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxyphenyl]methanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-methoxyethyl]-2-hydroxybenzenemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23NO3
CAS Number:
Molecular Weight:
253.34
Beilstein/REAXYS Number:
14479390
MDL number:
UNSPSC Code:
41116107

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

clinical testing

format

neat

SMILES string

OCC1=C(O)C=CC(C(OC)CNC(C)(C)C)=C1

InChI

1S/C14H23NO3/c1-14(2,3)15-8-13(18-4)10-5-6-12(17)11(7-10)9-16/h5-7,13,15-17H,8-9H2,1-4H3

InChI key

UMHASVFLCHGDPW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1B

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service