Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92675

Supelco

trans-2-Octenedioic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12O4
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

OC(CCCC/C=C/C(O)=O)=O

InChI

1S/C8H12O4/c9-7(10)5-3-1-2-4-6-8(11)12/h3,5H,1-2,4,6H2,(H,9,10)(H,11,12)

InChI key

BNTPVRGYUHJFHN-UHFFFAOYSA-N

Application

trans-2-Octenedioic acid may be used as an analytical reference standard for the quantification of the analyte in human urine samples using dual capillary column gas-liquid chromatography and gas-liquid chromatography coupled to mass spectrometry.[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of isomeric unsaturated medium-chain dicarboxylic acids in human urine
Jin JS and Tserng Y-K
Journal of Lipid Research, 30(10), 1611-1619 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service