Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92569

Supelco

Mexacarbate

PESTANAL®, analytical standard

Synonym(s):

4-Dimethylamino-3,5-dimethylphenyl N-methylcarbamate, 4-Dimethylamino-3,5-xylyl N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O2
CAS Number:
Molecular Weight:
222.28
Beilstein/REAXYS Number:
2216978
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C1=C(C)C=C(OC(NC)=O)C=C1C

InChI

1S/C12H18N2O2/c1-8-6-10(16-12(15)13-3)7-9(2)11(8)14(4)5/h6-7H,1-5H3,(H,13,15)

InChI key

YNEVBPNZHBAYOA-UHFFFAOYSA-N

General description

Mexacarbate belongs to the class of broad-spectrum insecticides, which are derived from carbamic acid.[1]

Application

Mexacarbate may be used as an analytical reference standard for the quantification of the analyte in environmental samples using chromatography techniques.[1][2]

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic method for the analysis of aminocarb, mexacarbate and some of their N-methylcarbamate metabolites by post-column derivatization with fluorescence detection
Sundaram, KMS and Curry, J
Journal of Chromatography A, 672(1-2), 117-124 (1994)
High-flow ion spray liquid chromatography/mass spectrometry
Hopfgartner, Gerard and Wachs, Timothy and Bean, Karen and Henion, Jack
Analytical Chemistry, 65(4), 439-446 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service