Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92499

Millipore

Triple Sugar Iron Agar (acc. to ISO)

NutriSelect® Plus, suitable for microbiology

Synonym(s):

TSI Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 12 g/L
dextrose, 1 g/L
ferric ammonium citrate, 0.3 g/L
lactose, 10 g/L
meat extract, 3 g/L
peptone, 20 g/L
phenol red, 0.025 g/L
sodium chloride, 5 g/L
sodium thiosulfate, 0.3 g/L
sucrose, 10 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

nonselective and differential for Citrobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for Proteus spp.
nonselective and differential for Salmonella spp.
nonselective and differential for Shigella spp.
nonselective and differential for coliforms
nonselective and differential for enterobacteriaceae

Application

Differential medium for identification of enterobacteria, according to ISO 6579, 6785 and 10272 Standards.

Preparation Note

Dissolve 64.6g of powder in 1 L of distilled water and bring to boiling. Dispense into tubes and sterilize at 121°C for 15 minutes. Leave to solidify with short slant and good butts.

Legal Information

NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service