Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92428

Supelco

Methylophiopogonanone A

analytical standard

Synonym(s):

(3R)-3-(1,3-Benzodioxol-5-ylmethyl)-2,3-dihydro-5,7-dihydroxy-6,8-dimethyl-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18O6
CAS Number:
Molecular Weight:
342.34
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

optical activity

[α]/D -75±5°, c = 0.1 in chloroform

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

−20°C

SMILES string

O=C1[C@H](CC2=CC=C(OCO3)C3=C2)COC4=C(C)C(O)=C(C)C(O)=C41

InChI

1S/C19H18O6/c1-9-16(20)10(2)19-15(17(9)21)18(22)12(7-23-19)5-11-3-4-13-14(6-11)25-8-24-13/h3-4,6,12,20-21H,5,7-8H2,1-2H3/t12-/m1/s1

InChI key

BXTNNJIQILYHJB-GFCCVEGCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service