Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92357

Supelco

Fenthion-ethyl

analytical standard

Synonym(s):

O,O-Diethyl O-(4-methylthio-m-tolyl) phosphorothioate, O,O-Diethyl O-[3-methyl-4-(methylthio)phenyl] phosphorothioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19O3PS2
CAS Number:
Molecular Weight:
306.38
Beilstein/REAXYS Number:
1985674
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

S=P(OC1=CC=C(SC)C(C)=C1)(OCC)OCC

InChI

1S/C12H19O3PS2/c1-5-13-16(17,14-6-2)15-11-7-8-12(18-4)10(3)9-11/h7-9H,5-6H2,1-4H3

InChI key

WGWJBWHBOKETRC-UHFFFAOYSA-N

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service