Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92223

Supelco

Theaflavin 3,3′-digallate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H32O20
CAS Number:
Molecular Weight:
868.70
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

Oc1cc(O)c2C[C@@H](OC(=O)c3cc(O)c(O)c(O)c3)[C@H](Oc2c1)C4=Cc5c(cc(O)c(O)c5C(=O)C(O)=C4)[C@H]6Oc7cc(O)cc(O)c7C[C@H]6OC(=O)c8cc(O)c(O)c(O)c8

InChI

1S/C43H32O20/c44-17-7-23(46)21-12-33(62-42(58)15-3-25(48)36(54)26(49)4-15)40(60-31(21)9-17)14-1-19-20(11-30(53)39(57)35(19)38(56)29(52)2-14)41-34(13-22-24(47)8-18(45)10-32(22)61-41)63-43(59)16-5-27(50)37(55)28(51)6-16/h1-11,33-34,40-41,44-51,53-55,57H,12-13H2,(H,52,56)/t33-,34-,40-,41-/m1/s1

InChI key

ZEASWHWETFMWCV-ISBUVJFSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Theaflavin 3,3′-digallate is a theaflavin derivative, exhibiting antioxidative, anti-inflammatory, and anti-tumor activities. It is also associated with many beneficial health effects, including the prevention of cancer and heart disease.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Theaflavin 3,3′-digallate may be used as an analytical reference standard for the quantification of the analyte in tea samples using high-performance liquid chromatography with tandem mass spectrometric detection.[2]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structural identification of mouse fecal metabolites of theaflavin 3, 3?-digallate using liquid chromatography tandem mass spectrometry
Chen H, et al.
Journal of Chromatography A, 1218(41), 7297-7306 (2011)
Simultaneous determination of eight catechins and four theaflavins in green, black and oolong tea using new HPLC?MS?MS method
Tao W, et al.
Journal of Pharmaceutical and Biomedical Analysis, 131(41), 140-145 (2016)
Susanne M Henning et al.
The Prostate, 75(5), 550-559 (2014-12-30)
Preclinical and epidemiologic studies suggest chemopreventive effects of green tea (GT) and black tea (BT) in prostate cancer. In the current study we determined the effect of GT and BT consumption on biomarkers related to prostate cancer development and progression.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service