Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92056

Supelco

Degalactotigonin

analytical standard

Synonym(s):

β-Lycotetraosyltigogenin, Dihydroaspidistrin, Uttronin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C50H82O22
CAS Number:
Molecular Weight:
1035.17
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥90.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@]12[C@](CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])C[C@@]5([H])[C@]4([H])[C@H](C)[C@]6(OC[C@H](C)CC6)O5)([H])C[C@@H](O[C@H]7[C@H](O)[C@@H](O)[C@@H](O[C@H]8[C@H](O[C@H]9[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O9)[C@@H](O[C@H]%10[C@H](O)[C@@H](O)[C@H](O)CO

InChI

1S/C50H82O22/c1-20-7-12-50(64-18-20)21(2)32-28(72-50)14-26-24-6-5-22-13-23(8-10-48(22,3)25(24)9-11-49(26,32)4)65-45-40(62)37(59)41(31(17-53)68-45)69-47-43(71-46-39(61)36(58)34(56)29(15-51)66-46)42(35(57)30(16-52)67-47)70-44-38(60)33(55)27(54)19-63-44/h20-47,51-62H,5-19H2,1-4H3/t20-,21+,22+,23+,24-,25+,26+,27-,28+,29-,30-,31-,32+,33+,34-,35-,36+,37-,38-,39-,40-,41+,42+,43-,44+,45-,46+,47+,48+,49+,50-/m1/s1

InChI key

BJNQXJIQCPPOHN-NUKVSUKUSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Frew et al.
Anticancer research, 14(6B), 2425-2428 (1994-11-01)
A convenient and reliable multisample assay for the screening of inhibitors of the growth factor signalling enzyme phosphatidylinositol-3-kinase (PtdIns-3-K) has been developed. Four natural product inhibitors of Ptdlns-3-K have been identified with IC50 values for hypericin 0.18 microM, emodin 3.3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service