Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91949

Supelco

5-Hydroxyomeprazole sulfide

analytical standard

Synonym(s):

4-Methoxy-6-{[(5-methoxy-1H-benzimidazol-2-yl)thio]methyl}-5-methyl-3-pyridinemethanol, 5-Methoxy-2-{[(5-hydroxymethyl-4-methoxy-3-methyl-2-pyridinyl)methyl]thio}-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3O3S
CAS Number:
Molecular Weight:
345.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc2[nH]c(SCc3ncc(CO)c(OC)c3C)nc2c1

InChI

1S/C17H19N3O3S/c1-10-15(18-7-11(8-21)16(10)23-3)9-24-17-19-13-5-4-12(22-2)6-14(13)20-17/h4-7,21H,8-9H2,1-3H3,(H,19,20)

InChI key

KBRHWXYFTPMNSX-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service