Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91938

Supelco

S-(1,2-Dicarboxyethyl)glutathione

≥95.0% (HPLC)

Synonym(s):

γ-Glu-Cys(succinic acid)-Gly, S-(2-Succinyl)glutathione, Lγ-Glutamyl-S-(1,2-dicarboxyethyl)-L-cysteinylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H21N3O10S
CAS Number:
Molecular Weight:
423.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

assay

≥95.0% (HPLC)

impurities

≤5.0% water

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CSC(CC(O)=O)C(O)=O)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C14H21N3O10S/c15-6(13(24)25)1-2-9(18)17-7(12(23)16-4-11(21)22)5-28-8(14(26)27)3-10(19)20/h6-8H,1-5,15H2,(H,16,23)(H,17,18)(H,19,20)(H,21,22)(H,24,25)(H,26,27)/t6-,7-,8?/m0/s1

InChI key

PWCIUOASSAHGHI-WPZUCAASSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

S-(2-Succinyl)glutathione is found in considerable amounts in rat liver, heart, and lens. It shows anti-inflammatory and anti-anaphylactic effects in vivo and inhibits histamine release from rat mast cells in vitro.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service