Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91903

Millipore

Bryant and Burkey Medium

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Bryant Burkey Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

beef extract, 7.5 g/L
casein enzymic hydrolysate, 15.0 g/L
L-cysteine hydrochloride, 0.50 g/L
resazurin, 0.0025 g/L
sodium acetate, 5.0 g/L
yeast extract, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

5.9±0.2 (25 °C)

application(s)

bioburden testing
food and beverages

microbiology

suitability

anaerobic bacteria
nonselective for Clostridium spp.
nonselective for bacteria (General Media)

Application

Bryant and Burkey Medium is used for detecting and enumerating spores of lactate fermenting Clostridium in milk and dairy products.

Preparation Note

Suspend 33 grams in 1000 ml distilled water containing 5 grams of Sodium lactate. Heat if necessary to dissolve the medium completely. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service