Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91742

Sigma-Aldrich

tert-Butyldimethylsilyl trifluoromethanesulfonate

purum, ≥98.0% (T)

Synonym(s):

TBDMS triflate, Trifluoromethanesulfonic acid tert-butyldimethylsilylester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Si(CH3)2C(CH3)3
CAS Number:
Molecular Weight:
264.34
Beilstein/REAXYS Number:
2370068
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

purum

assay

≥98.0% (T)

form

liquid

refractive index

n20/D 1.385 (lit.)

bp

65-67 °C/12 mmHg (lit.)

density

1.151 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)[Si](C)(C)OS(=O)(=O)C(F)(F)F

InChI

1S/C7H15F3O3SSi/c1-6(2,3)15(4,5)13-14(11,12)7(8,9)10/h1-5H3

InChI key

WLLIXJBWWFGEHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Powerful and strongly electrophilic reagent for the introduction of the tert-butyl-dimethylsilyl group; Reviews

replaced by

signalword

Danger

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

98.6 °F - closed cup

flash_point_c

37 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.V. Ley et al.
Tetrahedron, 55, 3515-3515 (1999)
E.J. Corey et al.
Journal of the American Chemical Society, 106, 3384-3384 (1984)
T. Mukaiyama et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 5, 121-121 (1999)
M. Riediker et al.
Helvetica Chimica Acta, 62, 205-205 (1979)
G. Simchen
Advances in Silicon Chemistry, 1 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service