Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91603

Millipore

Listeria mono Selective Supplement II

suitable for microbiology

Synonym(s):

Listeria monocytogenes Selective Supplement II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85
Product 91603 is not currently sold in your country. Contact Technical Service

agency

according to ISO 11290-1:2017
according to ISO 11290-2:2017

Quality Level

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

General description

A selective supplement recommended by ISO Committee for the isolation of Listeria species.

Application

for the isolation of Listeria species

Components

(per vial, sufficient for 500 ml medium)
Ceftazidime 10.0 mg
Nalidixic acid, sodium salt 10.0 mg
Amphotericin B 5.0 mg

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service