Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91569

Supelco

Strontium Standard for ICP

TraceCERT®, 10,000 mg/L Sr

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
Product 91569 is not currently sold in your country. Contact Technical Service

product line

TraceCERT®

Quality Level

grade

TraceCERT®

shelf life

limited shelf life, expiry date on the label

concentration

10,000 mg/L Sr

technique(s)

ICP: suitable

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Flame over circleCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 1 - Skin Corr. 1B

supp_hazards

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service