Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91566

Supelco

N-(Trimethylsilyl)acetamide

derivatization grade (GC derivatization), LiChropur, ≥98.0% (GC)

Synonym(s):

TMS acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHSi(CH3)3
CAS Number:
Molecular Weight:
131.25
Beilstein/REAXYS Number:
741928
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

derivatization grade (GC derivatization)

Quality Level

assay

≥98.0% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

bp

84 °C/18 mmHg (lit.)

mp

46-49 °C (lit.)

SMILES string

CC(=O)N[Si](C)(C)C

InChI

1S/C5H13NOSi/c1-5(7)6-8(2,3)4/h1-4H3,(H,6,7)

InChI key

LWFWUJCJKPUZLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of ascorbic acid.

Other Notes

Reagent for trimethylsilyl.

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

158.0 °F

flash_point_c

70 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M.Vecchi and K.Kaiser
Journal of Chromatography A, 26, 22-22 (1967)
D.R. Knapp
Handbook of Analytical Derivatization Reactions, 572-572 (1979)
Quantitative analysis of ascorbic acid in tissues by gas-liquid chromatography.
J H Allison et al.
Analytical biochemistry, 43(2), 401-409 (1971-10-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service