Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91287

Supelco

[10]-Shogaol

analytical standard

Synonym(s):

1-(4-Hydroxy-3-methoxyphenyl)-4-tetradecen-3-one, 10-Shogaol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H32O3
CAS Number:
Molecular Weight:
332.48
Beilstein/REAXYS Number:
20347696
UNSPSC Code:
85151701

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

form

solid

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

SMILES string

O=C(/C=C\CCCCCCCCC)CCC1=CC=C(O)C(OC)=C1

InChI key

FADFGCOCHHNRHF-VAWYXSNFSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service