Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

910422

Sigma-Aldrich

CX76

Umicore

Synonym(s):

Diacetatobis(tricyclohexylphosphine)palladium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H72O4P2Pd
CAS Number:
Molecular Weight:
785.36
UNSPSC Code:
12161600

form

powder

mp

211-216 °C

Application

Umicore CX76 is a homogeneous catalyst useful for cross coupling reactions with arenes, amination, Buchwald-Hartwig amination, Mizoroki-Heck coupling reactions, and telomerization.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service