Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909947

Sigma-Aldrich

3-Azido-L-alanine hydrochloride

≥95%

Synonym(s):

(S)-2-Amino-3-azidopropanoic acid hydrochloride, 3-Azido-L-alanine HCl, 3-Azido-L-alanine hydrochloride, H-L-Ala(N3)-OH*HCl, H-L-Aza-OH*HCl, H-L-Dap(N3)-OH*HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N4O2 · HCl
Molecular Weight:
166.57
UNSPSC Code:
12352209
Product 909947 is not currently sold in your country. Contact Technical Service

assay

≥95%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

storage temp.

2-8°C

Application

The in vivo incorporation of clickable unnatural amino acids such as 3-Azido-L-alanine with unique reactivity at a defined postition is used for functionalization of proteins. The azide functionalities in the protein can then be modified with almost any alkyne bearing molecule by Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC). Copper-free alternatives with strained internal alkynes are also available (SPAAC). Some examples enabled with this technique are protein PEGylation, masking with sugars, and the attachment to antibodies.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kristi L Kiick et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(1), 19-24 (2001-12-26)
The introduction of chemically unique groups into proteins by means of non-natural amino acids has numerous applications in protein engineering and functional studies. One method to achieve this involves the utilization of a non-natural amino acid by the cell's native

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service