Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909807

Sigma-Aldrich

4-Azido-D-homoalanine hydrochloride

≥98%

Synonym(s):

(R)-2-Amino-4-azidobutanoic acid hydrochloride, 4-Azido-D-homoalanine HCl, 4-Azido-D-homoalanine hydrochloride, H-D-Aha-OH*HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N4O2 · HCl
Molecular Weight:
180.59
UNSPSC Code:
12352202

assay

≥98%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

availability

available only in USA

storage temp.

2-8°C

Application

Some clickable unnatural D-amino acids can be fed to bacterial cells and incorporate into cell walls.The D-amino acids can also be used to generate L-RNA aptamers for the corresponding L-amino acid.

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Self-react. C

Storage Class

5.2 - Organic peroxides and self-reacting hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service