Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

909467

Sigma-Aldrich

Methyl 2,3,4,6-tetra-O-acetyl-β-D-glucopyranoside

≥95%

Synonym(s):

Methyl β-D-glucopyranoside tetraacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22O10
CAS Number:
Molecular Weight:
362.33
MDL number:

assay

≥95%

form

powder

storage temp.

2-8°C

InChI

1S/C15H22O10/c1-7(16)21-6-11-12(22-8(2)17)13(23-9(3)18)14(24-10(4)19)15(20-5)25-11/h11-15H,6H2,1-5H3

InChI key

UYWUMFGDPBMNCA-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service