Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90914

Supelco

AMCC

analytical standard

Synonym(s):

N-Acetyl-S-(N-methylcarbamoyl)cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2O4S
CAS Number:
Molecular Weight:
220.25
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

CNC(=O)SCC(NC(C)=O)C(O)=O

InChI

1S/C7H12N2O4S/c1-4(10)9-5(6(11)12)3-14-7(13)8-2/h5H,3H2,1-2H3,(H,8,13)(H,9,10)(H,11,12)

InChI key

MXRPNYMMDLFYDL-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Slatter et al.
Chemical research in toxicology, 4(2), 157-161 (1991-03-01)
S-(N-Methylcarbamoyl)-N-acetylcysteine (AMCC), a chemically labile mercapturic acid conjugate, was identified by liquid chromatography-mass spectrometry (LC-MS) in the urine of rats dosed intraperitoneally with methyl isocyanate (MIC; 45.2 mumol). The corresponding cysteine conjugate, however, was not detected in urine. Following methylation
A C Lareo et al.
International archives of occupational and environmental health, 67(1), 41-46 (1995-01-01)
Some methods for analysing N,N-dimethylformamide and its metabolites [hydroxymethyl-N-methylformamide, hydroxymethylformamide and N-acetyl-S-(N-methylcarbamoyl)cysteine] in the urine of exposed workers are described. Unchanged dimethylformamide was measured after pretreatment of urine (2 ml) with silica gel cartridges and elution with methanol. The gas
Swee Jin Tan et al.
PloS one, 8(7), e64084-e64084 (2013-07-31)
Library preparation for next-generation DNA sequencing (NGS) remains a key bottleneck in the sequencing process which can be relieved through improved automation and miniaturization. We describe a microfluidic device for automating laboratory protocols that require one or more column chromatography
J Mráz et al.
International archives of occupational and environmental health, 64(2), 85-92 (1992-01-01)
Excretion of N,N-dimethylformamide (DMF) and DMF metabolites N-hydroxymethyl-N-methylformamide ("MF"), N-hydroxymethyl-formamide ("F") and N-acetyl-S-(N-methylcarbamoyl)cysteine (AMCC) has been monitored in the urine of volunteers during and after their 8-h exposure to DMF vapour at a concentration of 10, 30 and 60 mg.m-3.
H U Käfferlein et al.
Journal of chromatography. B, Biomedical sciences and applications, 734(2), 285-298 (1999-12-14)
Two human urinary metabolites of the industrial solvent N,N-dimethylformamide (DMF), N-hydroxymethyl-N-methylformamide (HMMF) and N-acetyl-S-(N-methylcarbamoyl)cysteine (AMCC), were assayed using a new analytical method (gas chromatography and thermionic sensitive detection). Clean-up of urine samples includes a liquid-liquid extraction step followed by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service