Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

908878

Sigma-Aldrich

Chiralyst Rh1228

Umicore

Synonym(s):

(1,5-Cyclooctadiene)[(R)-1-[(R)-2-(2′-diphenylphosphinophenyl)ferrocenyl]-ethyldi-(bis-3,5-trifluoromethylphenyl)phosphine]rhodium(I) tetrafluoroborate, Chiralyst Rh1228

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H44BF16FeP2Rh
CAS Number:
Molecular Weight:
1228.41
UNSPSC Code:
12352101

form

powder or crystals

Application

Chiralyst Rh1228 is a homogeneous catalyst useful for asymmetric reactions, hydrogenation, asymmetric hydrogenation, and transfer hydrogenation.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Product License
This product, its manufacturing or use, is the subject of one or more issued or pending U.S. Patents (and foreign equivalents) owned or controlled by Umicore PMC. The purchase of this product from Umicore PMC through Sigma-Aldrich, its affiliates or their authorized distributors conveys to the buyer a limited, one-time, non-exclusive, non-transferable, non-assignable license. Buyer′s use of this product may infringe patents owned or controlled by third parties. It is the sole responsibility of buyer to ensure that its use of the product does not infringe the patent rights of third parties or exceed the scope of the license granted herein.

Additional information available at www.pmc.umicore.com

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service