907588
(4aR,6S,7R,8R,8aS)-7,8-bis(benzyloxy)-2,2-di-tert-butyl-6-(phenylthio)hexahydropyrano[3,2-d][1,3,2]dioxasiline
≥95%
Synonym(s):
Phenyl 2,3-O-dibenzyl-4,6-O-di-tert-butylsilyl-1-thio-β-D-galactoside
About This Item
Recommended Products
assay
≥95%
form
powder
mp
129-131 °C
storage temp.
2-8°C
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service