Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

907499

Sigma-Aldrich

(2R,3S,4S,5R,6S)-2-(((tert-Butyldimethylsilyl)oxy)methyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triol

≥95%

Synonym(s):

Phenyl 6-O-tert-butyldimethylsilyl-1-thio-β-D-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H30O5SSi
CAS Number:
Molecular Weight:
386.58
UNSPSC Code:
12352005

assay

≥95%

form

powder

mp

82-88 °C

storage temp.

2-8°C

Application

This is a protected glucopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has β-phenylthio and 6-tert-butyldimethylsilyl protecting groups.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service