This is a protected mannopyranoside useful as a building block for synthesis of complex carbohydrates. The compound has α-phenylthio, 3-benzyl and 4,6-benzylidene protecting groups.
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Carbohydrates and their biologically active glycoconjugate analogs are emerging as an important class of biomolecules.
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.