Skip to Content
MilliporeSigma

90710

2,2,2-Tribromoethanol

purum, ≥96.0% (GC)

Synonym(s):

Tribromoethyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Br3CCH2OH
CAS Number:
Molecular Weight:
282.76
EC Number:
200-903-1
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1733249
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥96.0% (GC)

bp

92-93 °C/10 mmHg (lit.)

mp

73-79 °C (lit.), 77-80 °C

SMILES string

OCC(Br)(Br)Br

InChI

1S/C2H3Br3O/c3-2(4,5)1-6/h6H,1H2

InChI key

YFDSDPIBEUFTMI-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library