Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90587

Supelco

Glucoraphenin potassium salt monohydrate

analytical standard

Synonym(s):

1-S-[5-(Methylsulfinyl)-N-(sulfooxy)-4-pentenimidoyl]-1-thio-β-D-glucopyranose monopotassium salt monohydrate, 4-(Methylsulfinyl)-3-butenylglucosinolate potassium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20KNO10S3 · H2O
CAS Number:
Molecular Weight:
491.60
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

analytical standard

form

solid

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service