Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90567

Supelco

Antipyrine

analytical reference material

Synonym(s):

2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Beilstein/REAXYS Number:
157775
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:

grade

analytical reference material
reference material

assay

≥98.5% (qNMR)
≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

109-111 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CN1N(C(=O)C=C1C)c2ccccc2

InChI

1S/C11H12N2O/c1-9-8-11(14)13(12(9)2)10-6-4-3-5-7-10/h3-8H,1-2H3

InChI key

VEQOALNAAJBPNY-UHFFFAOYSA-N

Gene Information

human ... PTGS1(5742)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L B Zavodnik et al.
Farmakologiia i toksikologiia, 52(3), 95-101 (1989-05-01)
The presented findings indicate that the determination in the blood or saliva of antipyrine concentration kinetics as well as the profile and the ratio of the products of its hydroxylation in the urine makes it possible to evaluate under clinical
Antipyrine as a model drug to study hepatic drug-metabolizing capacity.
H E Poulsen et al.
Journal of hepatology, 6(3), 374-382 (1988-06-01)
A P Viktorov et al.
Farmakologiia i toksikologiia, 53(1), 74-77 (1990-01-01)
The data about the use of "antipyrine test" in clinical practice for studying the drug oxidation peculiarities are presented. The data characterize the main pharmacokinetic properties of antipyrine as a "metabolic marker" for quantitative estimation of the effects of inductors
Factors influencing antipyrine elimination.
I H Stevenson
British journal of clinical pharmacology, 4(3), 261-265 (1977-06-01)
Measurement of blood flow with freely diffusible indicators as inert gases, antipyrine, labelled water and rubidium.
N A Lassen et al.
Acta endocrinologica. Supplementum, 158, 95-111 (1972-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service