Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90532

Supelco

N-Desisopropylpropranolol

analytical standard

Synonym(s):

(±)-1-Amino-3-(1-naphthyloxy)-2-propanol, (±)-Norpropranolol, N-Deisopropylpropranolol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15NO2
CAS Number:
Molecular Weight:
217.26
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water (calc. from elemental analysis)

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

NCC(O)COC1=CC=CC2=CC=CC=C21

InChI

1S/C13H15NO2/c14-8-11(15)9-16-13-7-3-5-10-4-1-2-6-12(10)13/h1-7,11,15H,8-9,14H2

InChI key

ZFMCITCRZXLMDJ-UHFFFAOYSA-N

General description

N-Desisopropylpropranolol is one of the active metabolites of propranolol, a widely prescribed β-adrenergic blocking drug for treating arrhythmias, thyrotoxicosis, angina pectoris, and hypertension.[1]

Application

N-Desisopropylpropranolol may be used as an analytical reference standard for the determination of the analyte in various biological samples[1][2][3][4] by chromatography techniques.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile isocratic high-performance liquid chromatographic assay for propranolol and its basic, neutral and acidic metabolites in biological fluids
Kwong EC and Shen DD
Journal of Chromatography. B, Biomedical Sciences and Applications, 414(65), 365-379 (1987)
Development and validation of a capillary zone electrophoresis method for the determination of propranolol and N-desisopropylpropranolol in human urine
Nevado JJB, et al.
Analytica Chimica Acta, 559(1), 9-14 (2006)
An improved HPLC-MS/MS method for simultaneous quantification of propranolol and its two phase I metabolites in plasma of infants with hemangioma and its application to a comparative study of plasma concentrations
He H, et al.
Royal Society of Chemistry Advances, 8(65), 37286-37294 (2018)
Rapid and simultaneous extraction of propranolol, its neutral and basic metabolites from plasma and assay by high-performance liquid chromatography
Harrison PM, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 343(1), 349-358 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service