Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

905135

Sigma-Aldrich

(±)-cis-N-Boc-2-fluorocyclohexylamine

≥95%

Synonym(s):

Monofluorinated ortho NBoc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H20FNO2
CAS Number:
Molecular Weight:
217.28
UNSPSC Code:
12352101

assay

≥95%

form

powder

storage temp.

2-8°C

Application

The incorporation of fluorine in agrochemical and pharmaceutical compounds is of high interest due to the ability to fine-tune the polarity, pKA-value, conformation and metabolic stability. Derived from a cis-selective hydrogenation developed by Glorius and coworkers, this fluorinated compound displays a diverse utility as a building block for chemical synthesis. Fluoro-cyclohexaneamine allows for the straightforward application in organic synthesis, after simple deprotection. It has been shown, that this compound can be used for the preparation of fluorinated analogs of Lomustine and Bromhexine.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mario P Wiesenfeldt et al.
Science (New York, N.Y.), 357(6354), 908-912 (2017-08-12)
All-cis-multifluorinated cycloalkanes exhibit intriguing electronic properties. In particular, they display extremely high dipole moments perpendicular to the aliphatic ring, making them highly desired motifs in material science. Very few such motifs have been prepared, as their syntheses require multistep sequences

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service