Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

903817

Sigma-Aldrich

cis-3,5-Difluoropiperidine

trifluoroacetate protected, ≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8F5NO
Molecular Weight:
217.14
UNSPSC Code:
12352101

assay

≥95%

form

powder

storage temp.

2-8°C

Application

Fluorinated piperidines are of interest in drugs, agrochemicals and materials science. A straightforward process developed by Glorius and coworkers allows the preparation of these motifs in a highly diastereoselective manner. The all-cis-3,5-difluoropiperidine can be directly employed in synthetic applications after TFA-deprotection.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zackaria Nairoukh et al.
Nature chemistry, 11(3), 264-270 (2019-01-22)
Piperidines and fluorine substituents are both independently indispensable components in pharmaceuticals, agrochemicals and materials. Logically, the incorporation of fluorine atoms into piperidine scaffolds is therefore an area of tremendous potential. However, synthetic approaches towards the formation of these architectures are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service