Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90331

Supelco

Arjunetin

analytical standard

Synonym(s):

(2α,3β,49α)- 2,3,19-Trihydroxyolean-12-en-28-oic acid β-D-glucopyranosyl ester, 24-Deoxysericoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H58O10
CAS Number:
Molecular Weight:
650.84
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@@H]1O)C(=O)O[C@@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O

InChI

1S/C36H58O10/c1-31(2)12-14-36(30(44)46-29-26(41)25(40)24(39)20(17-37)45-29)15-13-34(6)18(23(36)28(31)43)8-9-22-33(5)16-19(38)27(42)32(3,4)21(33)10-11-35(22,34)7/h8,19-29,37-43H,9-17H2,1-7H3/t19-,20-,21+,22-,23-,24-,25+,26-,27+,28+,29+,33+,34-,35-,36+/m1/s1

InChI key

CUOZRGBQTOSWAY-PWEPJYDOSA-N

Looking for similar products? Visit Product Comparison Guide

application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D V Singh et al.
Phytotherapy research : PTR, 18(2), 131-134 (2004-03-17)
Crude ethanolic extract of the stem bark of Terminalia arjuna (Combretaceae) and its three compounds namely arjunic acid, arjungenin and arjunetin were evaluated for antifeedant, growth inhibitory and oviposition-deterrent activities against a lepidopterous insect Spilarctia obliqua. The compound arjunetin showed
Ying Wang et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 43(5), 504-508 (2008-08-23)
To investigate the chemical constituents of the leaves of Adinandra nitida, several column chromatography methods were used to isolate the chemical constituents of this plant. The structures were elucidated on the basis of spectral data. Six compounds were isolated and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service