Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90299

Supelco

3-Methylpyrazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
Beilstein/REAXYS Number:
1454
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤3.0% water

refractive index

n20/D 1.491-1.497
n20/D 1.495 (lit.)

bp

204 °C (lit.)

density

1.02 g/mL at 25 °C (lit.)

application(s)

environmental

SMILES string

Cc1cc[nH]n1

InChI

1S/C4H6N2/c1-4-2-3-5-6-4/h2-3H,1H3,(H,5,6)

InChI key

XKVUYEYANWFIJX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service