Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

900646

Sigma-Aldrich

1,2-Dimethoxyethane

anhydrous, inhibitor-free, ZerO2®, 99.5%

Synonym(s):

1,2-Dimethoxyethane ZerO2®, Degassed and low oxygen 1,2-dimethoxyethane, mono-Glyme, Dimethylglycol, Ethylene glycol dimethyl ether, Monoglyme

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OCH3
CAS Number:
Molecular Weight:
90.12
Beilstein/REAXYS Number:
1209237
EC Number:
MDL number:
UNSPSC Code:
12352112
grade:
ZerO2®
assay:
99.5%
bp:
85 °C (lit.)
vapor pressure:
48 mmHg ( 20 °C)

grade

ZerO2®

Quality Level

vapor density

3.1 (20 °C, vs air)

vapor pressure

48 mmHg ( 20 °C)

assay

99.5%

form

liquid

autoignition temp.

396 °F

quality

anhydrous

expl. lim.

10.4 %

dilution

(for analytical testing)

refractive index

n20/D 1.379 (lit.)

bp

85 °C (lit.)

mp

−58 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

SMILES string

COCCOC

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

Inchi Key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

41.0 °F - closed cup

flash_point_c

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service