Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

90009

Supelco

3,4-Dimethyl-1H-pyrazole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
Beilstein/REAXYS Number:
1621
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental

format

neat

SMILES string

Cc1c[nH]nc1C

InChI

1S/C5H8N2/c1-4-3-6-7-5(4)2/h3H,1-2H3,(H,6,7)

InChI key

VQTVFIMEENGCJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dimethyl-1H-pyrazole is a substituted pyrazole compound, which has a five-membered ring containing two heteroatoms as the active constituent.[1][2]

Application

3,4-Dimethyl-1H-pyrazole may be used as an analytical standard for the determination of the analyte in environmental samples and Onobrychis spp. pollens by liquid chromatography-tandem mass spectrometry (LC-MS/MS) and gas chromatography-mass spectrometry (GC-MS) techniques.[3][4]

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid analysis of the nitrification inhibitor 3, 4-dimethylpyrazole phosphate in soil using LC-MS/MS
Doran GS, et al.
International Journal of Environmental Analytical Chemistry, 98(7), 606-621 (2018)
Novel synthesis of the nitrification inhibitor 3, 4-dimethyl-1H-pyrazole phosphate
Chen Z, et al.
J. Chem. Res. (M), 37(11), 652-653 (2013)
Gas chromatography and mass spectrometry analysis, chemical contents and antioxidant properties of Onobrychis spp.(Fabaceae) pollen collected by honeybees
Temizer IK, et al.
Fresenius Environmental Bulletin, 26(1 A), 962-968 (2017)
Product class 1: pyrazoles
Stanovnik B and Svete J
Sci. Synth., 12, 15-225 (2002)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service