Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89960

Supelco

Ginkgotoxin

analytical standard

Synonym(s):

4′-O-Methylpyridoxine, 4′-Methoxypyridoxine, 4-(Methoxymethyl)pyridoxine, 5-Hydroxy-3-(hydroxymethyl)-4-(methoxymethyl)-6-methylpyridine, 5-Hydroxy-4-(methoxymethyl)-6-methyl-3-pyridinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO3
CAS Number:
Molecular Weight:
183.20
Beilstein/REAXYS Number:
150788
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

OCC1=CN=C(C)C(O)=C1COC

InChI

1S/C9H13NO3/c1-6-9(12)8(5-13-2)7(4-11)3-10-6/h3,11-12H,4-5H2,1-2H3

InChI key

SVINQHQHARVZFF-UHFFFAOYSA-N

General description

Ginkgotoxin is an alkaloid, which occurs in the seeds and leaves of Ginkgo biloba.[1][2] It is also a potential antivitamin, structurally related to vitamin B6.[2]

Application

Ginkgotoxin may be used as a standard in the determination of ginkgotoxin in selected Chinese herbs and plants using high-performance liquid chromatography (HPLC) or gas chromatography coupled with mass spectrometry (GC-MS).[3]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The human pyridoxal kinase, a plausible target for ginkgotoxin from Ginkgo biloba.
Kastner U, et al.
FEBS Journal, 274(4), 1036-1045 (2007)
Extraction and chromatography-mass spectrometric analysis of the active principles from selected Chinese herbs and other medicinal plants.
Wang X, et al.
American Journal of Chinese Medicine, 31(06), 927-944 (2003)
Medical Toxicology of Natural Substances: Foods, Fungi, Medicinal Herbs, Plants, and Venomous Animals.
Jones ID and Slovis CM
JAMA : The Journal of the American Medical Association, 274(4), 1036-1045 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service