Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89927

Supelco

L-Carnitine-(trimethyl-13C3) inner salt hydrate

analytical standard

Synonym(s):

(R)-3-Carboxy-2-hydroxy-N,N,N-trimethyl-13C3-1-propanaminium inner salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C3C4H15NO3 · xH2O
Molecular Weight:
164.18 (anhydrous basis)
UNSPSC Code:
41116107
NACRES:
NB.21

grade

analytical standard

assay

≥97.0% (TLC)

optical activity

[α]/D -32.0±3.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

extent of labeling

≥99.0% (LC-MS)

impurities

≤15% water

format

neat

storage temp.

2-8°C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service