89926
(2-Hydroxyethyl-d4)phosphonic acid
PESTANAL®, analytical standard
Synonym(s):
2-Hydroxyethane-d4-phosphonic acid, Ethephon-hydroxy-d4
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
assay
≥98.0% (GC)
shelf life
limited shelf life, expiry date on the label
mp
52-60 °C
application(s)
agriculture
format
neat
storage temp.
2-8°C
General description
Application
Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1C
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service