Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89925

Supelco

Valeramide

pharmaceutical impurity standard

Synonym(s):

Valproic acid impurity E (PhEur), NSC 400223, Pentanamide, Valeric acid amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO
CAS Number:
Molecular Weight:
101.15
Beilstein/REAXYS Number:
1740792
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

SMILES string

NC(CCCC)=O

InChI

1S/C5H11NO/c1-2-3-4-5(6)7/h2-4H2,1H3,(H2,6,7)

InChI key

IPWFJLQDVFKJDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhang et al.
Food chemistry, 339, 128014-128014 (2020-11-07)
The research of host insect-resistance is a hot spot in the field of controlling storage pests. However, the mechanism of host resistance to storage pests is still unclear. Some researchers think it is related to grain cracking characters of rice
Kevin A Kam et al.
Sensors (Basel, Switzerland), 18(9) (2018-08-26)
We have pioneered the use of liquid polar organic molecules as alternatives to rigid gate-dielectrics for the fabrication of graphene field-effect transistors. The unique high net dipole moment of various polar organic molecules allows for easy manipulation of graphene's conductivity
Dinisha Cyril Pirapaharan et al.
Calcified tissue international, 104(1), 92-101 (2018-09-09)
Cathepsin K (CatK) inhibition allows reducing bone resorption with specific advantages compared to the existing anti-osteoporosis drugs. Its clinical use appears even more promising with the recent development of ectosteric inhibitors. A confusing observation, however, is that a low dose
Claire Villette et al.
Scientific reports, 8(1), 13178-13178 (2018-09-06)
Barley (Hordeum vulgare) is the fourth crop cultivated in the world for human consumption and animal feed, making it important to breed healthy and productive plants. Among the threats for barley are lodging, diseases, and pathogens. To avoid lodging, dwarf

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service