Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

89885

Supelco

Ritalinic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17NO2
CAS Number:
Molecular Weight:
219.28
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

shipped in

wet ice

storage temp.

2-8°C

SMILES string

OC(=O)C(C1CCCCN1)c2ccccc2

InChI

1S/C13H17NO2/c15-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-14-11/h1-3,6-7,11-12,14H,4-5,8-9H2,(H,15,16)

InChI key

INGSNVSERUZOAK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olof Beck et al.
Therapeutic drug monitoring, 36(4), 528-534 (2014-01-24)
It has been discovered recently that exogenous substances are detectable in exhaled breath after intake. Exhaled breath therefore constitutes a new possible matrix in clinical pharmacology and toxicology. The present work was aimed at exploring this possibility further by a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service