Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3CH=C(CH3)COOH
CAS Number:
Molecular Weight:
100.12
EC Number:
201-295-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
1236500
MDL number:
grade
purum
assay
≥97.0% (GC)
bp
95-96 °C/12 mmHg (lit.)
mp
61-64 °C
density
0.969 g/mL at 25 °C (lit.)
SMILES string
C\C=C(/C)C(O)=O
InChI
1S/C5H8O2/c1-3-4(2)5(6)7/h3H,1-2H3,(H,6,7)/b4-3+
InChI key
UIERETOOQGIECD-ONEGZZNKSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.