89392
Sinensetin
analytical standard
Synonym(s):
2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxy-4H-1-benzopyran-4-one, 2-(3,4-Dimethoxyphenyl)-5,6,7-trimethoxychromen-4-one, 3′,4′,5,6,7-Pentamethoxyflavone, 6-Hydroxyluteolin 5,6,7,3′,4′-pentamethyl ether, Pedalitin permethyl ether
About This Item
Recommended Products
grade
analytical standard
assay
≥95.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
≤5.0% water
application(s)
food and beverages
format
neat
storage temp.
2-8°C
SMILES string
O=C1C=C(C2=CC(OC)=C(OC)C=C2)OC3=C1C(OC)=C(OC)C(OC)=C3
InChI
1S/C20H20O7/c1-22-13-7-6-11(8-15(13)23-2)14-9-12(21)18-16(27-14)10-17(24-3)19(25-4)20(18)26-5/h6-10H,1-5H3
InChI key
LKMNXYDUQXAUCZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Recommended products
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service