Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89242

Supelco

2-Methoxyestradiol

analytical standard

Synonym(s):

1,3,5(10)-Estratriene-2,3,17-triol 2-methyl ether, 2,3,17β-Trihydroxy-1,3,5(10)-estratriene 2-methyl ether, 2-Hydroxyestradiol 2-methyl ether, 3,17β-Dihydroxy-2-methoxy-1,3,5(10)-estratriene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O3
CAS Number:
Molecular Weight:
302.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

OC1=C(C=C(C2=C1)[C@]3([H])[C@](CC2)([H])[C@]4([H])CC[C@H](O)[C@](C)4CC3)OC

InChI

1S/C19H26O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,18,20-21H,3-8H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1

InChI key

CQOQDQWUFQDJMK-SSTWWWIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B T Zhu et al.
Cancer research, 58(11), 2269-2277 (1998-06-11)
Catechol estrogens (2- or 4-hydroxyestradiol and 2- or 4-hydroxyestrone) are chemically reactive estrogen metabolites that are O-methylated to less polar monomethyl ethers by catechol-O-methyltransferase, an enzyme present in many tissues such as the liver, kidney, brain, placenta, uterus, and mammary
Low catechol-O-methyltransferase and 2-methoxyestradiol in preeclampsia: more than a unifying hypothesis.
Duska Dragun et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 24(1), 31-33 (2008-11-29)
G Schumacher et al.
Journal of cancer research and clinical oncology, 127(7), 405-410 (2001-07-27)
2-Methoxyestradiol (2-ME) is a physiological metabolite of estrogen, which is excreted with the urine. In contrast to most estrogens, tumor growth-inhibiting effects were observed. Further studies have revealed that it may be an effective anticancer compound for many tumor types.
Jean-François Peyrat et al.
Current medicinal chemistry, 19(24), 4142-4156 (2012-06-20)
2-Methoxyestradiol (2ME2), a natural metabolite of estradiol which has no estrogenic activity, is a potent antitumor and anti-angiogenic compound, currently undergoing clinical trials for treatment of a variety of cancers. In the last two decades, an ever increasing number of
Svetlana Verenich et al.
Molecular pharmaceutics, 7(6), 2030-2039 (2010-09-14)
2-Methoxyestradiol (2MeO-E2) is an endogenous metabolite of estrogen which was initially considered to be inactive. During the last few decades it has been shown that 2MeO-E2 is a promising anticancer drug. In vitro experiments have demonstrated that it has several

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service