Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89032

Sigma-Aldrich

Thioredoxin from Spirulina sp.

≥60% (GE)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202

form

powder

concentration

≥60% (GE)
≥60% protein

solubility

Tris pH 8.0: 2 mg/mL, very slightly hazy, yellow

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mediates the reduction of disulfide bonds in proteins.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin A Turturice et al.
PLoS pathogens, 9(1), e1003136-e1003136 (2013-02-06)
The Plasmodium ookinete develops over several hours in the bloodmeal of its mosquito vector where it is exposed to exogenous stresses, including cytotoxic reactive oxygen species (ROS). How the parasite adapts to these challenging conditions is not well understood. We
Franco L Simonetti et al.
Nucleic acids research, 41(Web Server issue), W8-14 (2013-05-30)
MISTIC (mutual information server to infer coevolution) is a web server for graphical representation of the information contained within a MSA (multiple sequence alignment) and a complete analysis tool for Mutual Information networks in protein families. The server outputs a
Anna Lenart et al.
Biochimica et biophysica acta, 1834(7), 1279-1284 (2013-04-02)
The small, obscure group of selenoprotein oxidoreductases and the huge clan of kinases, the workhorses of cellular signalling, are rarely discussed together. Focusing on selenoproteins of unknown structures, we predict a thioredoxin-like fold for the Selenoprotein N (SelN) family and
Effects of thioredoxin-1 on neurogenesis after brain ischemia/reperfusion injury.
Feng Zhou et al.
CNS neuroscience & therapeutics, 19(3), 204-205 (2013-02-28)
Takashi Ashino et al.
Arteriosclerosis, thrombosis, and vascular biology, 33(4), 760-768 (2013-02-16)
Reactive oxygen species are important mediators for platelet-derived growth factor (PDGF) signaling in vascular smooth muscle cells, whereas excess reactive oxygen species-induced oxidative stress contributes to the development and progression of vascular diseases, such as atherosclerosis. Activation of the redox-sensitive

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service