88885
Thiomorpholine
purum, ≥97.0% (GC)
Synonym(s):
Tetrahydro-2H-1,4-thiazine, Thiamorpholine
About This Item
grade
purum
assay
≥97.0% (GC)
refractive index
n20/D 1.538 (lit.)
n20/D 1.540
bp
169 °C (lit.)
density
1.026 g/mL at 25 °C (lit.)
SMILES string
C1CSCCN1
InChI
1S/C4H9NS/c1-3-6-4-2-5-1/h5H,1-4H2
InChI key
BRNULMACUQOKMR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
signalword
Danger
hcodes
Hazard Classifications
Skin Corr. 1B
Storage Class
8A - Combustible corrosive hazardous materials
wgk_germany
WGK 3
flash_point_f
145.4 °F - closed cup
flash_point_c
63 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service