Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88823

Supelco

N-(7-Nitro-4-benzofurazanyl)-D-prolyl chloride

for fluorescence

Synonym(s):

(R)-(+)-1-(7-Nitro-4-benzofurazanyl)-2-pyrrolidinecarbonyl chloride, (R)-(+)-4-(2-Chlorformyl-pyrrolidino)-7-nitrobenzofurazan, (R)-(+)-NBD-Pro-COCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9ClN4O4
CAS Number:
Molecular Weight:
296.67
MDL number:
UNSPSC Code:
23151817

grade

for fluorescence

form

powder

fluorescence

λex 484 nm; λem 528 nm in acetonitrile

suitability

suitable for fluorescence

storage temp.

2-8°C

InChI

1S/C11H9ClN4O4/c12-11(17)8-2-1-5-15(8)6-3-4-7(16(18)19)10-9(6)13-20-14-10/h3-4,8H,1-2,5H2

InChI key

XBQYXPYABMCADZ-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Fluorescent reagent for the resolution of alcohols and amines by HPLC[1]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T. Toyo-oka et al.
Journal of Chromatography A, 675, 79-79 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service