Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88300

Supelco

2-Thenoyltrifluoroacetone

for spectrophotometric det. of metal ions, ≥99.0%

Synonym(s):

4,4,4-Trifluoro-1-(2-thienyl)-1,3-butanedione, TTA (extractant), TTB, TTFA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5F3O2S
CAS Number:
Molecular Weight:
222.18
Beilstein/REAXYS Number:
168645
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥99.0% (GC)
≥99.0%

form

crystals

quality

for spectrophotometric det. of metal ions

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

UV/Vis spectroscopy: suitable

bp

96-98 °C/8 mmHg (lit.)

mp

40-44 °C (lit.)
42-46 °C

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)CC(=O)c1cccs1

InChI

1S/C8H5F3O2S/c9-8(10,11)7(13)4-5(12)6-2-1-3-14-6/h1-3H,4H2

InChI key

TXBBUSUXYMIVOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Thenoyltrifluoroacetone may be used as a chelating agent for the speciation of chromium in water and rice samples using surfactant-assisted emulsification dispersive liquid-liquid microextraction with electrothermal atomic absorption spectrometry technique.[1]

Other Notes

Reagent for the extraction and spectrophotometric determination of metals[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

233.6 °F - closed cup

flash_point_c

112 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z. Holzbecher et al.
Handbook of Organic Reagents in Inorganic Analysis (1976)
Surfactant-assisted emulsification dispersive liquid-liquid microextraction using 2-thenoyltrifluoroacetone as a chelating agent coupled with electrothermal atomic absorption spectrometry for the speciation of chromium in water and rice samples
Dokpikul N, et al.
Food Chemistry, 246(1-2), 379-385 (2018)
Feng Gao et al.
Talanta, 80(1), 202-206 (2009-09-29)
Amino-functionalized luminescent silica nanoparticles (LSNPs) doped with the europium(III) mixed complex, Eu(TTA)(3)phen with 2-thenoyltrifluoroacetone (TTA) and 1,10-phenanthroline(phen) were synthesized successfully using an revised Stöber method. Scanning electron microscopy (SEM), transmission electron microscopy (TEM), Fourier-transform infrared (FTIR), and fluorescence spectroscopy were
David González-Aragón et al.
Biochemical pharmacology, 73(3), 427-439 (2006-11-25)
Dicoumarol, a competitive inhibitor of NAD(P)H:quinone oxidoreductase 1 (NQO1), increases intracellular superoxide and affects cell growth of tumor cells. This work was set to establish a mechanistic link between dicoumarol, superoxide and cell cycle alterations in HL-60 cells. Using ES936
Tao Huang et al.
Journal of cellular biochemistry, 107(5), 973-983 (2009-06-06)
Gap junctional intercellular communication (GJIC) plays a critical role in the control of multiple cell behavior as well as in the maintenance of tissue and organ homeostasis. However, mechanisms involved in the regulation of gap junctions (GJs) have not been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service