Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88060

Supelco

Tetraphenylphosphonium chloride

for the spectrophotometric det. of Bi, Co, ≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4P(Cl)
CAS Number:
Molecular Weight:
374.84
Beilstein/REAXYS Number:
3922393
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

assay

≥97.0% (AT)
≥97.0%

form

crystals

quality

for the spectrophotometric det. of Bi, Co

technique(s)

UV/Vis spectroscopy: suitable

mp

272-274 °C (lit.)

SMILES string

[Cl-].c1ccc(cc1)[P+](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20P.ClH/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;1H/q+1;/p-1

InChI key

WAGFXJQAIZNSEQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Zhou et al.
Bioconjugate chemistry, 22(8), 1459-1472 (2011-06-24)
Alteration in mitochondrial transmembrane potential (ΔΨ(m)) is an important characteristic of cancer. The observation that the enhanced negative mitochondrial potential is prevalent in tumor cell phenotype provides a conceptual basis for development of mitochondrion-targeting therapeutic drugs and molecular imaging probes.
Do Kyun Kim et al.
Journal of microbiology and biotechnology, 23(3), 430-435 (2013-03-07)
Multidrug resistance, especially multidrug efflux mechanisms that extrude structurally unrelated cytotoxic compounds from the cell by multidrug transporters, is a serious problem and one of the main reasons for the failure of therapeutic treatment of infections by pathogenic microorganisms as
Myocardial imaging for mitochondrial membrane potential.
H William Strauss et al.
JACC. Cardiovascular imaging, 5(3), 293-296 (2012-03-17)
Denice C Bay et al.
Biochimica et biophysica acta, 1798(3), 526-535 (2009-12-29)
Escherichia coli multidrug resistance protein E (EmrE) is a four transmembrane alpha-helix protein, and a member of the small multidrug resistance protein family that confers resistance to a broad range of quaternary cation compounds (QCC) via proton motive force. The
Yael Elbaz et al.
The Journal of biological chemistry, 283(18), 12276-12283 (2008-03-07)
Glycine residues may play functional and structural roles in membrane proteins. In this work we studied the role of glycine residues in EmrE, a small multidrug transporter from Escherichia coli. EmrE extrudes various drugs across the plasma membrane in exchange

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service