Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87994

Supelco

Tetraoctylammonium bromide

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7]4N(Br)
CAS Number:
Molecular Weight:
546.79
Beilstein/REAXYS Number:
4169176
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

description

for ion-selective electrodes

Quality Level

product line

Selectophore

form

crystals

mp

95-98 °C (lit.)
98-100 °C

SMILES string

[Br-].CCCCCCCC[N+](CCCCCCCC)(CCCCCCCC)CCCCCCCC

InChI

1S/C32H68N.BrH/c1-5-9-13-17-21-25-29-33(30-26-22-18-14-10-6-2,31-27-23-19-15-11-7-3)32-28-24-20-16-12-8-4;/h5-32H2,1-4H3;1H/q+1;/p-1

InChI key

QBVXKDJEZKEASM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Tetraoctylammonium bromide (TOAB) is an ionophore or recognition element mostly used in potentiometric sensors.
Visit our Sensor Applications portal to learn more.

Application

TOAB has been used as recognition element in formulating potentiometric membrane to develop an electronic tongue system for monitoring nitrogen species level in water samples.[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

213.8 °F - closed cup

flash_point_c

101 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of plasticizer on the selectivity of nitrate-sensitive CHEMFETs
W.P.R.V. Stauthamer, J.F.J. Engbersen, W. Verboom, D.N. Reinhoudt
Sensors and Actuators B, Chemical, 17, 197-201 (1994)
Development and application of an electronic tongue for detection and monitoring of nitrate, nitrite and ammonium levels in waters.
Nu?ez, L., et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 110, 273-279 (2013)
Lin Han et al.
Dalton transactions (Cambridge, England : 2003), 43(31), 11981-11987 (2014-06-28)
Phase transfer techniques possess remarkable advantages for the synthesis of inorganic nanomaterials. In contrast to the abundant reports on the preparation of noble metal nanoparticles using phase transfer, the number of semiconductor nanocrystals syntheses based on phase transfer techniques is
Yu Cheng et al.
Small (Weinheim an der Bergstrasse, Germany), 10(24), 5137-5150 (2014-08-12)
The blood-brain barrier (BBB) remains a formidable obstacle in medicine, preventing efficient penetration of chemotherapeutic and diagnostic agents to malignant gliomas. Here, a transactivator of transcription (TAT) peptide-modified gold nanoparticle platform (TAT-Au NP) with a 5 nm core size is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service